Great Plains Industries Heavy-Duty Turbine Flowmeter/Totalizer Accessories
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
05609-91 05609-92
MFLX05609-91EA 144 EUR InStock
MFLX05609-91 MFLX05609-92
Great Plains Industries Heavy-Duty Turbine Flowmeter/Totalizer Accessories
Durchflussmesser
Optimize and enhance performance of your Masterflex flowmeter.
  • A wide range of accessories are available to provide further functionality to your flowmeter
These accessories are ideal for the Masterflex® Heavy-Duty Turbine Flowmeters/Totalizers. Choode from the optional remote installation kit, replacement battery kits, output modules, and calibration containers to enhance performance of your flowmeters.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge