Surfactants, Tergitol™ 15-S-7
Lieferant: Spectrum Chemical
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
T1258-100ML T1258-4LT T1258-500ML
SPCMT1258-100MLEA 101 EUR InStock
SPCMT1258-100ML SPCMT1258-4LT SPCMT1258-500ML
Surfactants, Tergitol™ 15-S-7
Reinigungsmittel Waschmittel und Oberflächenreiniger
Tergitol™ 15-S-7, is a non-ionic Secondary Alcohol Ethoxylate surfactant and is an excellent emulsifier and detergent, enhances paint thickener efficiency and aids in rinseability of solvent-based systems. This product is suggested for use in cleaners, pre-wash spotters and paints and coatings.
  • Superior wetting and rinseability
  • Excellent formulation and handling properties
  • Low odour
  • Narrow gel range
  • Soluble in water
Density at 20 °C (68 °F): 0,991 g/ml
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge