AP biology investigation 5: cell processes: photosynthesis, Ward's®
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
WARD470134-770EA 316 EUR InStock
WARD470134-770
AP biology investigation 5: cell processes: photosynthesis, Ward's®
Meets AP science practices 1, 2, 3, 6, and 7.
  • Updated illustrated instructions aid comprehension
  • Leaf pigment separation and environmental effects on photosynthetic oxygen production is quantified
  • Available for individual student use or class-size kit
In this investigation, the ability of plants to capture, store, and use energy from light for growth and reproduction is brought to life as students practice basic paper chromatography techniques to visualize pigments present in various plant leaves. Using a floating disk assay, students will investigate the environmental elements that affect the rate of photosynthesis under controlled conditions.
The Only Kits Aligned with AP Science Practices. Ward's AP Biology Investigations provide complete coverage of the Four Big Ideas and Seven Science Practices to help you meet new AP Biology curriculum standards and prepare students for real-world success in college and research-level labs. Each investigation follows a series of three parts, beginning with a structured lab investigation, followed by a related guided inquiry investigation with thoughtful assessment questions, and finally, suggestions to encourage students to formulate their own questions and explore further self-guided experimentation. This complete series of investigations emphasizes the progression to fully student-directed investigations and builds competence in all the science practices, helping students:
• Practice concept application instead of fact memorization.
• Focus on self-guided experimentation instead of step-by-step lab procedure.
• Investigate real-world concepts with greater curriculum flexibility.
• Apply problem-solving skills to strengthen student inquiry.
AP and Advanced Placement Program are registered trademarks of the College Entrance Examination Board, which was not involved in the production of and does not endorse these products.
Individual student kit contains reduced quantities of materials to perform most activities, but on an individual student basis.
Zubehörinformationen: Individual student kit contains reduced quantities of materials to perform most activities, but on an individual student basis.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge