Benzo[1,2-b:4,5-b']dithiophene-4,8-dione 97%
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
BD226528-25G BD226528-100G
BLDPBD226528-25GEA 366 EUR InStock
BLDPBD226528-25G BLDPBD226528-100G
Benzo[1,2-b:4,5-b']dithiophene-4,8-dione 97%
Benzo[1,2-b:4,5-b']dithiophene-4,8-dione
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel: C₁₀H₄O₂S₂
Storage Temperature: Raumtemperatur
MDL Number: MFCD01927240
CAS-Nummer: 32281-36-0
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge