VWR®, Method 1613 precision and recovery std native compounds, varied conc, in nonane, CRM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
91606.001EA 5040 EUR InStock
91606.001 91605.010
VWR®, Method 1613 precision and recovery std native compounds, varied conc, in nonane, CRM
Standards Organische Standards
  • 17 components
  • Varied concentration in Nonane
Analytes (target concentration):
2,3,7,8-Tetrachlorodibenzo-p-dioxin (CAS 1746-01-6) 40 ng/mL
2,3,7,8-Tetrachlorodibenzofuran (CAS 51207-31-9) 40 ng/mL
1,2,3,7,8-Pentachlorodibenzo-p-dioxin (CAS 40321-76-4) 200 ng/mL
1,2,3,7,8-Pentachlorodibenzofuran (CAS 57117-41-6) 200 ng/mL
2,3,4,7,8-Pentachlorodibenzofuran (CAS 57117-31-4) 200 ng/mL
1,2,3,4,7,8-Hexachlorodibenzo-p-dioxin (CAS 39227-28-6) 200 ng/mL
1,2,3,6,7,8-Hexachlorodibenzo-p-dioxin (CAS 57653-85-7) 200 ng/mL
1,2,3,7,8,9-Hexachlorodibenzo-p-dioxin (CAS 19408-74-3) 200 ng/mL
1,2,3,4,7,8-Hexachlorodibenzofuran (CAS 55684-94-1) 200 ng/mL
1,2,3,6,7,8-Hexachlorodibenzofuran (CAS 57117-44-9) 200 ng/mL
1,2,3,7,8,9-Hexachlorodibenzofuran (CAS 72918-21-9) 200 ng/mL
2,3,4,6,7,8-Hexachlorodibenzofuran (CAS 60851-34-5) 200 ng/mL
1,2,3,4,6,7,8-Heptachlorodibenzo-p-dioxin (CAS 35822-46-9) 200 ng/mL
1,2,3,4,6,7,8-Heptachlorodibenzofuran (CAS 38998-75-3) 200 ng/mL
1,2,3,4,7,8,9-Heptachlorodibenzofuran (CAS 55673-89-7) 200 ng/mL
1,2,3,4,6,7,8,9-Octachlorodibenzo-p-dioxin (CAS 3268-87-9) 400 ng/mL
1,2,3,4,6,7,8,9-Octachlorodibenzofuran (CAS 39001-02-0) 400 ng/mL
Zertifizierungen: Certified reference material.
Achtung! Bitte beachten: Flammable.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge