VWR®, ICP-OES Wavelength calibration solution, 15 components in 5% HNO₃
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
91216.260EA 563 EUR InStock
91216.260 91216.180
VWR®, ICP-OES Wavelength calibration solution, 15 components in 5% HNO₃
Standards Kationischer Standards Mehrelement-Standards
15 components: Al 5 ug/ml, As 5 ug/ml, Ba 5 ug/ml, Cd 5 ug/ml, Co 5 ug/ml, Cr 5 ug/ml, Cu 5 ug/ml, Mn 5 ug/ml, Mo 5 ug/ml, Ni 5 ug/ml, Pb 5 ug/ml, Se 5 ug/ml, Sr 5 ug/ml, Zn 5 ug/ml, K 50 ug/ml in 5% HNO₃
Zertifizierungen: ISO 17025/17034
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge