DermaLife™ K Keratinocyte Medium Kit
Lieferant: Lifeline Cell Technology
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
MSPPLL-0007EA 0 EUR InStock
MSPPLL-0007
DermaLife™ K Keratinocyte Medium Kit
Zellkulturmedien
DermaLife™ K has been optimised for the culture of Human Epidermal Keratinocytes in a serum-free environment.
  • Contains supplements and growth factors
  • Grows human epidermal keratinocytes
  • Maintains excellent cell morphology
  • Contains no phenol red or antimicrobials
DermaLife™ K LifeFactors® Kit contains supplements and growth factors for use with human keratinocytes and DermaLife basal medium. DermaLife™ K Medium grows human epidermal keratinocytes through at least 15 population doublings at rates that meet or exceed other commercially available serum-free media while maintaining excellent cell morphology. DermaLife™ K contains no phenol red or antimicrobials.
Lifeline® optimised culture media contain no antimicrobials and no phenol red.
Bestellinformation: DermaLife™ K Medium Complete Kit includes DermaLife basal medium and DermaLife K LifeFactors® Kit of associated supplements and growth factors. The LifeFactors® include L-Glutamine 15 ml, Extract P® 2 ml, Epinephrine 0,5 ml, rh TGF-α 0,5 ml, Hydrocortisone Hemisuccinate 0,5 ml, rh Insulin 0,5 ml, Apo-Transferrin 0,5 ml, Calcium Chloride 0,5 ml, Gentamicin and Amphotericin B 0,5 ml.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge