Alkaline Phosphatase (APP) Chromogen Substates for IHC
Lieferant: VECTOR LABORATORIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
SK-5100 SK-5200 SK-5300 SK-5400 SK-5105
VECTSK-5100EA 470 EUR InStock
VECTSK-5100 VECTSK-5200 VECTSK-5300 VECTSK-5400 VECTSK-5105
Alkaline Phosphatase (APP) Chromogen Substates for IHC
Elektrophorese-Reagenzien Western Blotting Reagents
Vector Laboratories alkaline phosphatase (AP) substrate options include standard substrates and ImmPACT substrates with higher sensitivity. The range of vibrant colors facilitates multiplex staining and improved contrast with pigmented tissues and counterstains.
  • Optimised for IHC, ICC, ISH and blots
  • Stock solutions are supplied in convenient dropper bottles for greater ease of handling
  • Compatible with either VectaMount Permanent Mounting Media (H-6000) or VectaMount AQ Aqueous Mounting Media (H-5501)
  • One year expiry date
For immuno-visualisation experiments to be successful, one needs proper sample prep, a well-targeted primary anybody, an effective secondary antibody-based detection system and a high-quality chromogenic substrate. Vector Laboratories has decades of experience offering exceptional staining systems and substrates.
Standard substrates provide exceptional value and convenience over substrates in the form of powders or pills and work well for higher-abundance targets. Vector Laboratories ImmPACT substrates offer up to five times higher sensitivity with convenient handling and are ideal for detecting lower-abundance targets.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge