Lactose, wasserfrei NF
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
LA103-12KG LA103-2.5KG LA103-500GM
SPCMLA103-12KGEA 1740 EUR InStock
SPCMLA103-12KG SPCMLA103-2.5KG SPCMLA103-500GM
Lactose, wasserfrei NF
Laktose
Anhydrous Lactose, NF is used as a tablet diluent in the pharmaceutical industry.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formel: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Siedepunkt: 592 °C (1013 hPa)
Melting Pt: 222.8 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00151251
CAS-Nummer: 63-42-3
EINECS: 200-559-2
Merck Index: 12,05356
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge