Matte Amber Ultra Heat Resistant Polyimide Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
174187 174375 173776 174341 173725 174215 173865 174253 173884 173723 173779 173699 173774
BRDY174187EA 211.68 EUR InStock
BRDY174187 BRDY174375 BRDY173776 BRDY174341 BRDY173725 BRDY174215 BRDY173865 BRDY174253 BRDY173884 BRDY173723 BRDY173779 BRDY173699 BRDY173774
Matte Amber Ultra Heat Resistant Polyimide Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Etiketten
Amber high temperature polyimide with an ultra-durable adhesive withstands the extreme wash protocol and cleaning chemicals used in printed circuit board manufacturing.
  • Withstands multiple cycles of harsh condition washes
  • Resists high temperatures up to 300 °C (572 °F)
  • Survives on the top or bottom side of printed circuit boards through the solder process
  • Matte finish
Ideal for circuit board labelling and component and equipment labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Zertifizierungen: Meets the performance requirements of MIL -STD-202G, Notice 12, Method 215K. Meets the requirements of SAE AS81531 Marking of Electrical Insulating Material.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge