Self-Laminating Self-Extinguishing Tedlar Wrap Around Label for M710 and BMP71
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
173375 173394 173339
BRDY173375EA 70.35 EUR InStock
BRDY173375 BRDY173394 BRDY173339
Self-Laminating Self-Extinguishing Tedlar Wrap Around Label for M710 and BMP71
Etiketten
The B-642 translucent polyvinylflouride material with permanent adhesive has excellent flexibility for wrapping and small diameter wires.
  • Self-extinguishing material is ideal for wire and cable marking applications in aerospace, defence, and mass transit industries
  • Excellent solvent resistance
  • Self-laminating
  • Matte finish
Ideal for data and telecommunications labelling, and wire and cable labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R4300.
Zertifizierungen: RoHS compatibility Pass 2011/65/EU.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge