Flame-Resistant Standard Coats
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
FR34NARL FR34NARM FR34NARS FR34NARXL FR34NARXXL FR34NARXXXL
PRTWFR34NARLEA 48.4 EUR InStock
PRTWFR34NARL PRTWFR34NARM PRTWFR34NARS PRTWFR34NARXL PRTWFR34NARXXL PRTWFR34NARXXXL
Flame-Resistant Standard Coats
Arbeitskleidung
Flame-resistant coats are designed to be worn over flame-resistant garments. Constructed with a comfortable, lightweight, highly innovative flame-resistant twill fabric.
  • Three pockets for ample storage
  • Pen division on left pocket
  • Concealed stud front for easy access
  • Lightweight and comfortable
  • 0+ UPF rated fabric to block 98% of UV rays
  • CE-Cat. III
FR standard coats provide protection against radiant, convective, and contact heat. The lightweight design enhances wearer comfort.
Zertifizierungen: EN ISO 11612 A1+A2, B1, C1, E2, F1. EN ISO 11611 Class 1 A1+A2. EN 1149 -5. ASTM F1959/F1959M-14e1 ATPV 9.1 cal/cm². CE certified.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge