Safety Storage Cabinet, Type 90, ION-CORE-90 STORE model IO90.195.060.CS.WDC
Lieferant: asecos
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
MSGA41362EA 190.5 EUR InStock
MSGA41362 135-2883 MSGA41363 MSGA41364 MSGA41365
Safety Storage Cabinet, Type 90, ION-CORE-90 STORE model IO90.195.060.CS.WDC
Möbel Lagerschränke
Safe passive storage of lithium-ion batteries.
  • Robust construction and longevity
  • Integrated potential-free alarm contact
  • No unauthorised use
  • Integrated transport base
  • Easy alignment
  • Smoke detector
  • Pressure relief
  • All-around protection
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface. Integrated potential-free alarm contact for strongly recommended alarm forwarding. Optionally available with module for remote signalling.
No unauthorised use: door lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green). Permanent locking of the door by automatic 2-point locking. Integrated transport base: In the event of an emergency, immediate evacuation, and transport of the cabinet to a safe place outdoors.
Easy alignment: adjusting aids to compensate for uneven floor. Smoke detector for fire detection and forwarding the alarm to the building management system. Pressure relief in the head section.
All-around protection: 90 minutes fire protection from outside to inside (type 90/type tested in accordance with EN 14470-1) and for more than 90 minutes fire resistance for fires from inside to outside.
Zertifizierungen: Fire resistance 90 minutes. Type tested in accordance with EN 14470-1. CE compliant.
Bestellinformation: Interior equipment packages need to be ordered separately (not included). Please contact your Avantor representative for more information and available options.
Lieferumfang: Delivery prices available upon request.
Achtung! Bitte beachten: According to the security concept it is highly recommended to place ION-LINE cabinets at the ground level in a location from where a quick evacuation from the building is possible in the event of fire.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge