Retro Reflective All-Weather Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
170809 170810 170811 170812 170935 170936 170937 170938
BRDY170809EA 41.23 EUR InStock
BRDY170809 BRDY170810 BRDY170811 BRDY170812 BRDY170935 BRDY170936 BRDY170937 BRDY170938
Retro Reflective All-Weather Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Etiketten
Increase safety and visibility with the B-584 highly-reflective label material. Print labels almost anywhere high visibility is needed with a compact Brady handheld label printer.
  • Reflective
  • Outdoor durable
  • Abrasion resistant
  • Low temperature resistant
  • Heat resistant
  • Gloss finish
Ideal for safety applications such as on personnel, fire suppression equipment, and first aid cabinets or anywhere extra visibility is required. Resistant to abrasions, weather, solvents and extreme temperatures. Pressure-sensitive acrylic adhesive withstands prolonged exposure to tough conditions. Able to withstand outdoor conditions for 4 to 6 years on average.
Gloss finish, abrasion-resistant, chemical-resistant, outdoor use B-584 labels are compatible with Brady printer models M410, BMP41, M510, BMP51, BMP53, and M511.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge