Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Laboratory Freezers, ZLN-T 300

Lieferant: POL-EKO APARATURA SP.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

ZLNT300PSMPR ZLNT300PSSMPR
POLEZLNT300PSMPREA 5440 EUR InStock
POLEZLNT300PSMPR POLEZLNT300PSSMPR
Laboratory Freezers, ZLN-T 300
Gefriergeräte Gefriergeräte für das Labor und allgemeine Anwendungen
Laboratory freezers ZLN-T 300 temperature range of −40 to 0°C and a user-friendly interface. Conduct precise experiments with quality control at −40 °C, and multilingual support. Stay connected with LAN and USB ports, enhance security with door locks and alarms. The Smart Pro edition boasts WiFi communication and LabDesk software.

  • Quality control protocol (at −20 °C)
  • Instruction manual in English and menu in various languages
  • Open door alarm
  • Units have castors as standard
  • LAN port and USB port
  • Access port (Ø 20 mm, on the left side) for an external sensor
  • Door lock
  • Full solid shelves with a central hole
  • Full external doors
  • WiFi communication (Smart Pro only)
  • LAN cable (Smart Pro only)
  • LabDesk software for, amongst other things, downloading data to computer (Smart Pro only, optional for Smart version)
  • Natural air convection with working capacity 262 L

Elevate your laboratory operations with the ZLN-T 300 laboratory freezer, designed for excellence. With an impressive temperature range of −40 to 0 °C, this freezer enables meticulous experiments, including quality control at −40 °C.

Its user-friendly interface, multilingual support, and connectivity features like LAN and USB ports ensure seamless functionality. Security is prioritised with door locks, alarms, and a quality control protocol at −20 °C. The Smart Pro edition takes innovation a step further, offering WiFi communication and LabDesk software for efficient data management.

With castors for mobility, a door lock for security, and full external doors for accessibility, this freezer stands at the pinnacle of laboratory equipment.

Whether you opt for the Smart Pro version with additional features like WiFi communication and LabDesk software or the Smart version with optional upgrades, the ZLN-T 300 is a symbol of cutting-edge technology in laboratory freezers. The environmental friendly refrigerant (R290) has a very low global warming potential (GWP is three).

Lieferumfang: Supplied with two full solid shelves with a central hole. Other accessories are not included and must be ordered separately. User manual can be downloaded from controller, quick start guide supplied in chamber.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge