Sulphite Water Test Kit
Lieferant: TINTOMETER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
56P018640 56L018765
LOVI56P018640EA 20.8 EUR InStock
LOVI56P018640 LOVI56L018765
Sulphite Water Test Kit
Umwelt-Prüfkits
Provides accurate and reliable water analysis solutions that are valuable in a range of industries.
  • Colours may vary depending on sample and test conditions
  • Ignore any undissolved material after powder/tablet addition
  • For concentrations of sodium sulphite above 150 mg/l take a 10 ml sample and use a factor of 10 (i.e. each drop of Sulphite Titrant S2 used = 10 mg/l Na2SO₃)
Catalysed sulphite reacts quickly with atmospheric oxygen when hot, so the sample should be cooled during collection with the minimum of contact with air. It should be tested immediately after it has cooled. Care should be taken when obtaining samples.
Sulphite reserve may be expressed in different ways. To convert readings from sodium sulphite multiply the result obtained by the following factors. Sodium sulphite to sodium metabisulphite×0,8 and sodium sulphite to sulphite×0,63.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge