Labels for BBP33 Printers, Type B-777
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
153291 153287 153286 153284 153290 153282 153283
BRDY153291EA 143.65 EUR InStock
BRDY153291 BRDY153287 BRDY153286 BRDY153284 BRDY153290 BRDY153282 BRDY153283
Labels for BBP33 Printers, Type B-777
Etiketten
Printed circuit board and electronic component pre-process identification running smoothly with B-777 Polyimide labels.
  • Glossy white non-reflow circuit board labels
  • Service temperature of –70 to 300 °C
  • Suitable for outdoor use
  • Rectangle with rounded corners
This material does not require reflow or preheating to ensure printing readability, making it ideal for boards that go through multiple harsh-condition washes.
Use in combination with B30-R6300 ribbon to meet MIL-STD-202G, Method 215K resistance to solvents test requirements.
Use of B30-R6000 ribbon is acceptable but B30-R6300 is recommended for best performance in washdown cycles - user should test to their intended application.
Abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant and weather-resistant.
Zertifizierungen: Compliant with Directive 2011/65/EU and 2015/863.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge