Labels for BBP33 Printers, B-413
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
153136 153135 153134 152871 153132 153139 153133 153138 153137
BRDY153136EA 230.03 EUR InStock
BRDY153136 BRDY153135 BRDY153134 BRDY152871 BRDY153132 BRDY153139 BRDY153133 BRDY153138 BRDY153137
Labels for BBP33 Printers, B-413
Etiketten
Non-metal polyester (B-413) is a thermal transfer polyester material with a metallic-looking finish. Ideal for barcoded asset tracking and rating plate applications.
  • Light grey top-coated material with permanent adhesive adheres to most clean, dry surfaces
  • Thermal transfer printable
  • Service temperature of ‒56 to +43 °C
  • Abrasion-, chemical-, corrosion- and humidity-resistant
A ribbon is required to print this material. Used in barcode labeling, component and equipment labelling and general identification.
Zertifizierungen: UL approved. Compliant with Directive 2011/65/EU and 2015/863.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge