Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Laboratory Refrigerators, CHL 500

Lieferant: POL-EKO APARATURA SP.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

CHL500PMSMPR CHL500PMSSMPR
POLECHL500PMSMPREA 4320 EUR InStock
POLECHL500PMSMPR POLECHL500PMSSMPR
Laboratory Refrigerators, CHL 500
Kühlschränke
CHL Laboratory Refrigerators feature a cooling system to maintain temperatures from 0 to 15 °C, these specialised units prioritise stability and uniformity. Equipped with advanced microprocessor temperature controllers: Smart or Smart PRO, and forced air circulation, POL-EKO refrigerators guarantee precise temperature control for accredited laboratories, ensuring the utmost sample protection.

  • Quality control protocol (at +4 °C)
  • Instruction manual in English and menu in various languages
  • Over temperature protection 1.0 class (Smart) and 3.2 class (Smart PRO) according to DIN 12880
  • Monoblock cooling unit
  • Open door alarm, door lock
  • LAN port and USB port, internal LED light
  • Access port (Ø 30 mm, on the left side) for an external sensor
  • Stainless steel wire shelves for C (comfort) and P (premium) models
  • Solid external doors, assembly kit for fixing to the wall
  • WiFi communication (Smart Pro only), LAN cable (Smart Pro only)
  • LabDesk software for, amongst other things, downloading data to computer (Smart Pro only, optional for Smart version)

The CHL 500 refrigerators are more than just cold storage; they are equipped with forced air circulation for optimal temperature uniformity. The advanced microprocessor temperature controller ensures stability and uniformity, and its large, colourful display allows intuitive operation even when wearing gloves. Unlike traditional refrigerators, the compressor activates as needed, prioritising temperature control.

The two-component polyurethane foam insulation and eco-friendly refrigerants contribute to energy efficiency and environmental sustainability and the monoblock cooling unit contributes to optimal performance. The environmentally friendly refrigerant (R290) has a very low global warming potential (GWP is 3). 469 L working capacity.

Security is paramount, with features such as over-temperature protection, open door alarms, and a secure door lock. The internal LED lighting enhances visibility, and the access port accommodates an external sensor for added monitoring capabilities. The CHL 500 refrigerators are available as standard with solid doors or with glass doors (as a factory fitted option), providing options to suit your specific storage needs.

Customise your CHL refrigerator further with optional add-ons, including WiFi communication, LAN ports, and LabDesk software. This integration enables enhanced communication and data management, providing a streamlined and connected laboratory experience.

Bestellinformation: For all units operating at set point ≤5 °C the automatic defrosting function (POLEST/CHLPLUS) must be ordered to prevent icing.

Lieferumfang: Supplied with 3 stainless steel wire shelves. Other options and accessories are not included and must be ordered separately. User manual can be downloaded from controller, quick start guide supplied in chamber.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge