Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Cooled Incubators, ST 1450

Lieferant: POL-EKO APARATURA SP.

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

ST1450PMSMARTP ST1450PMSSMPRO
POLEST1450PMSMARTPEA 7060 EUR InStock
POLEST1450PMSMARTP POLEST1450PMSSMPRO
Cooled Incubators, ST 1450
Brutschränke Kühlbrutschränke
ST 1450 Cooled Incubators ensure reliable temperature control from 3 to 70 °C, regardless of ambient conditions. POL-EKO's models boast internal LED lighting, secure lockable doors. Ideal for stability testing and pharmaceutical storage, these incubators offer versatile chamber configurations, optimising laboratory space and enabling independent temperature control in each compartment.

  • Quality control protocol (at 37 °C)
  • Instruction manual in English and menu in various languages
  • Over temperature protection 1.0 class (according to DIN 12880) for C (comfort) models, 2.0 for P (premium) models and 3.3 class in Smart PRO
  • Monoblock cooling unit
  • Castors
  • Assembly kit for fixing to the wall
  • Open door alarm; Door lock; Solid external doors
  • LAN port and USB port; Access port (Ø30 mm, on the left side) for an external sensor
  • Internal LED light
  • Stainless steel wire shelves for C (comfort) and P (premium) models
  • Wi-Fi communication and LAN cable (Smart Pro only)
  • LabDesk software for, amongst other things, downloading data to computer (Smart Pro only, this is optional for Smart version)

Elevate your laboratory capabilities with the ST 1450 Cooled Incubator COMFORT or PREMIUM models, featuring a SMART/SMART PRO controller for precision. Providing a stabile temperature range from 3 to 40 °C and forced air convection, these incubators guarantee reliable sample storage and incubation. The 1525 L chamber capacity, three stainless steel wire shelves, and a 30 mm access port provide ample space and flexibility.

Benefit from features like USB port, automatic defrosting, and interior LED lighting. The solid door, door lock, and monoblock cooling unit contribute to optimal performance. The 24-month warranty adds peace of mind. Explore the Smart Pro options for enhanced communication, including WiFi, LAN port, and LabDesk software.

Discover unparalleled control with the ST 1450 Cooled Incubators. The solid door, stainless steel wire shelves, and access port enhance usability. Security features like over-temperature protection, open door alarm, and door lock guarantee a controlled environment. Benefit from additional features such as LAN and USB ports, and optional Smart Pro communication capabilities. Trust POL-EKO for reliable, efficient, and technologically advanced cooled incubators. The environmentally friendly refrigerant (R290) has a very low global warming potential (GWP is 3).

Lieferumfang: Supplied with 2×3 stainless steel wire shelves. Other accessories are not included and must be ordered separately. User manual can be downloaded from controller, quick start guide supplied in chamber.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge