PW377 - PW3 Class 1 Zipped Winter Hoodies
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
PW377OBR4XL PW377OBRL PW377OBRM PW377OBRS PW377OBRXL PW377OBRXXL PW377OBRXXXL
PRTWPW377OBR4XLEA 69.9 EUR InStock
PRTWPW377OBR4XL PRTWPW377OBRL PRTWPW377OBRM PRTWPW377OBRS PRTWPW377OBRXL PRTWPW377OBRXXL PRTWPW377OBRXXXL
PW377 - PW3 Class 1 Zipped Winter Hoodies
Arbeitskleidung
The PW3 Class 1 zipped winter hoodie is characterised by its distinctive design, combining Portwest HiVisTex Pro reflective tape with contemporary contrast panels. Offering exceptional comfort and warmth, 2 ample side zipped pockets, the fit is roomy and the fabric is soft to touch.
  • Knitted fabric with brushed backing
  • Fleece lining for added warmth and comfort
  • Lightweight flexible HiVisTex Pro segmented reflective tape for increased visibility
  • Two side zip pockets for secure storage
  • Hood for added protection against the elements
  • Ribbed hem for a comfortable fit
  • Ribbed cuffs for added comfort
  • Heat applied segmented reflective tape for added visibility
  • 40+ UPF rated fabric to block 98% of UV rays
Portwest PW3™ contemporary workwear blends sports and lifestyle trends with function, a modern fit and improved comfort. Utilising premium fabrics, products are engineered for flexibility across a diverse range of industry and trade end uses and offer exceptional value.
The extensive range of high-visibility clothing meets stringent requirements in both design and construction, to ensure compliance with the latest EN ISO 20471 and ANSI standards. Innovative and technical, our High Visibility range is ideal for those who will not compromise on style, comfort, protection and performance.
Shell fabric: 65% polyester, 35% cotton 300 g
Lining fabric: 100% polyester borg fleece 300 g
Zertifizierungen: Certified to EN ISO 20471 after 50× washes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge