PharmaFluor® FEP Tubings
Lieferant: SAINT GOBAIN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
PHFE6037506250 PHFE6037503150
GESSPHFE6037506250EA 575 EUR InStock
GESSPHFE6037506250 GESSPHFE6037503150
PharmaFluor® FEP Tubings
Schläuche Transferschlauch
PharmaFluor® FEP fluoropolymer tubing meets the highest demands of biopharmaceutical, medical, and laboratory applications. This tubing is formulated to resist the absorption/adsorption of aqueous fluids, which minimises the risk of fluid alteration in single- or repeat- use applications. PharmaFluor FEP has outstanding chemical and physical characteristics and can be used in a broad range of temperatures, from −73 to +204 °C (−100 to +400 °F).
  • Chemically inert
  • Ultra-pure
  • Documented biocompatibility
  • Fully characterised
  • Extremely low sorption characteristics
  • Fully sterilisable via autoclave or gas
  • Excellent barrier with very low permeability
  • Autoclavable
PharmaFluor® FEP tubing has outstanding chemical and physical characteristics, and processing of PharmaFluor® FEP occurs under stringently controlled conditions. In addition to being chemically inert, this tubing can be used at temperatures ranging from −73 to +204 °C (−100 to +400 °F). It is also virtually unaffected by chemical sanitisers and cleaners and can be cleaned repeatedly without decreasing its service life. It is ideal for applications such as sterile filling and dispensing systems, insulin production, diagnostic equipment, general laboratory use, cell and tissue culture transport, analytical and process equipment in the biotechnology, pharmaceutical and environmental fields.
Hardness: 55 (shore D)
Zertifizierungen: USP Class VI criteria, ISO 10993 and European Pharmacopoeia 3.1.9.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge