BradyJet™ J2000 Safety Marine Labels, White
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
150758 150759
BRDY150758EA 182.93 EUR InStock
BRDY150758 BRDY150759
BradyJet™ J2000 Safety Marine Labels, White
Etiketten
The British standard BS 5609 sets durability criteria for printed pressure-sensitive, adhesive-coated labels used when shipping chemical drums by sea.
  • Keep it colorful - this material runs through the BradyBS 5609 section 2 and 3 compliant Jet J2000 for high-resolution full-color printing
  • Keep it compliant - these labels are certified for use in global marine transport and are ideal for GHS chemical labels
  • BradyJet printers let you create the full-color labels you need to meet your customer and in-house specifications
  • Inkjet printing lets you create the full-color labels you need without the adhesive build up that can break down laser printers
  • An ink cartridge is required to print this material
Application: GHS and chemical labels, indoor/outdoor use, hazard communication identification, marine labelling.
Zertifizierungen: BS 5609 section 2 and 3 compliant. Compliant with Directive 2011/65/EU and 2015/863.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge