xGen™ ssDNA and Low-Input DNA Library Prep Kit
Lieferant: INTEGRATED DNA TECHNOLOGIE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
10009817 10009859
IDNA10009817EA 5310 EUR InStock
IDNA10009817 IDNA10009859
xGen™ ssDNA and Low-Input DNA Library Prep Kit
Nukleinsäure-Reagenzien Next Generation Sequencing Reagenzien
The xGen ssDNA and low-input DNA library prep kit enables preparation of libraries from degraded and damaged ssDNA and dsDNA samples in a single reaction. This kit allows users to sequence difficult-to-process samples via unique, proprietary adaptaseTM technology.
  • Low input, high complexity - inputs as low as 10 pg from highly degraded samples
  • Compatible with ssDNA and dsDNA samples - preserves input fragmentation patterns for precise mapping of DNA insert ends
  • Simple two-hour protocol
  • Workflow design for easy automation
Library prep kit for ssDNA (heavily damaged) or mixtures of ss and dsDNA (viromes/metagenomes) from 10 pg to 250 ng. Adaptase™-based chemistry for library prep of single stranded DNA, includes amplification polymerase.
Minimise sequence bias from PCR with high-fidelity PCR polymerase. Unique chemistry adapts short DNA fragments ≥40 bp long. Multiplex up to 1536 samples - pre-plated UDI primers enable multiplexing of up to 1536 samples. Compatible with xGen normalase™ technology - enzymatic normalisation of libraries from a single pool of libraries.
Lieferumfang: Indexing primers also supplied separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..