Brady® Chemical-Resistant Cryogenic Polypropylene Labels for M410, M510, and M511 Printers
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
177013 177012
BRDY177013EA 61.71 EUR InStock
BRDY177013 BRDY177012
Brady® Chemical-Resistant Cryogenic Polypropylene Labels for M410, M510, and M511 Printers
Etiketten
Read text and scan barcodes easily with high-performance B-7425 chemical- and temperature-resistant polypropylene, which is designed to resist harsh laboratory processes.
  • Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces like glass, copper, and stainless steel
  • Compatible with Brady printers BMP41, BMP51, BMP53, M410, M510, and M511
  • For use with ribbon series R4300
Brady's B-7425 chemical- and temperature-resistant polypropylene labels outlast extremes and are ideal for laboratory identification on vials, centrifuge tubes, and test tubes. These labels resist chemicals, liquid nitrogen, and other harsh environments while offering a high/low temperature range of −80 to +70 °C.
Zertifizierungen: RoHS compliant to 2011/65/EU (Pass 2015/863).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge