Natriumalginat (aus Macrocystis pyrifera (kelp)), Sigma-Aldrich®
Lieferant: SIGMAALDRICH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
M-Clarity™ Qualitätsstufe = MQ200
A2033-100G A2033-1KG A2033-250G A2033-500G
SIALA2033-100GEA 128 EUR InStock
SIALA2033-100G SIALA2033-1KG SIALA2033-250G SIALA2033-500G
Natriumalginat (aus Macrocystis pyrifera (kelp)), Sigma-Aldrich®
Natriumalginat
Alginate, a colloidal polyuronic acid structural molecule capable of gelation, is used in the preparation of colloidal biodegradable structures such as gels, biofilms, beads, nanoparticles, and microcapsules suitable for applications that range from gel based separation technologies to drug delivery and cell preservation.
  • Medium viscosity, ≥2,000 cP
Melting Pt: >300 °C
Storage Temperature: Kühlschrank
MDL Number: MFCD00081310
CAS-Nummer: 9005-38-3
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge