ESD Safety Sandals, Velcro®, Ian XXTP Red Easy Low S1
Lieferant: ELTEN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
711310/36 711310/37 711310/38 711310/39 711310/35 711310/40 711310/41 711310/42 711310/43 711310/44 711310/45 711310/46 711310/47 711310/48 711310/49
ELTN711310/36EA 97.9 EUR InStock
ELTN711310/36 ELTN711310/37 ELTN711310/38 ELTN711310/39 ELTN711310/35 ELTN711310/40 ELTN711310/41 ELTN711310/42 ELTN711310/43 ELTN711310/44 ELTN711310/45 ELTN711310/46 ELTN711310/47 ELTN711310/48 ELTN711310/49
ESD Safety Sandals, Velcro®, Ian XXTP Red Easy Low S1
Schuhe und Stiefel
ESD sandals made from microfibre/mesh upperl with fabric lining and a TPU/PU sole (Wellmaxx Trainers Power).
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Antistatic soft fleece insole
  • Abrasion-proof toe protection
  • Composite toe cap
Ideal for use in dry work areas, industry, storage, transport, assembly etc. (S1), areas where there is a risk of electrostatic discharge (ESDS/ESD). E.g. airports, airplane construction, automobile manufacturing.
Colour: Red
Zertifizierungen: EN ISO 20345 S1 FO/SR/SC, form A.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge