Digital Pressure Probes with Tube Port
Lieferant: Greisinger
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
486677 486675 486674 486676 486678 486679
GRSG486677EA 0 EUR InStock
GRSG486677 GRSG486675 GRSG486674 GRSG486676 GRSG486678 GRSG486679
Digital Pressure Probes with Tube Port
Temperaturfühler
DX 210 and DX 240 are series of miniature digital absolute and differential pressure probes for the PRO D01/PRO D05 handheld meters.
  • Interchangeable
  • Low power
  • Flexible hose connector
  • Compact and rigid stainless steel body
  • Fast and accurate
  • Temperature compensated
  • Optional DAkkS/ACCREDIA certification available
The measurementis temperature compensated for enhanced accuracy. The probes are supplied factory-calibrated, and they are ready to use. The calibration data are stored in the probe, allowing for interchangeability without the need for recalibration. Equipped with 4-pin M12 connector 'DX-Sensor standard', connectable to PRO-D0x multifunctional meters.
Bestellinformation: They can be optionally supplied with a DAkkS/ACCREDIA calibration certificate.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge