Corning® CoolCell® LX Cell Freezing Containers
Lieferant: Stemcell Technologies
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
200-0644 200-0643 200-0642 200-0645
STMC200-0644EA 468 EUR InStock
STMC200-0644 STMC200-0643 STMC200-0642 STMC200-0645
Corning® CoolCell® LX Cell Freezing Containers
Boxen Kühl- und Gefrierboxen, Kühl- und Gefrierbeutel
Ensure standardised freezing - without the use of alcohol - for most cell types with Corning® CoolCell® LX Cell freezing containers.
  • Eliminate the need for isopropanol with alcohol-free cryopreservation
  • Obtain reproducible freezing profiles by using controlled-rate freezing
  • Non-absorbent foam construction protects neighboring frozen samples and minimise impacts to the freezer environment
  • Rapidly equilibrate to room temperature upon removal from freezer
  • Easily identify samples with numbered wells
This container achieves consistent freezing profiles by combining a solid state core, radial vial symmetry, and uniform-density, cross-linked polyethylene foam. The polyethylene foam is non-absorbent, imposing negligible change to the freezer environment and protecting nearby samples. Use Corning® CoolCell® LX in a –80 °C freezer to achieve reproducible freezing at the rate of –1 °C/minute, which is ideal for the cryopreservation of most cells and cell lines. Each freezing container can hold 12 standard 1 to 2 ml cryogenic vials.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge