Brady® UL Approved Polyester Labels for BradyJet™ J7300 Colour Label Printer
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
176599 176595 176598 176591 176594 176597 176596 176592 176593
BRDY176599EA 439.63 EUR InStock
BRDY176599 BRDY176595 BRDY176598 BRDY176591 BRDY176594 BRDY176597 BRDY176596 BRDY176592 BRDY176593
Brady® UL Approved Polyester Labels for BradyJet™ J7300 Colour Label Printer
Etiketten
Polyester material meets UL 969 standard for marking and labelling and is ideal for product labelling, logos, 'Right to Know' labels, and barcode applications.
  • Durable and resistant to abrasions, chemicals, oils, and solvents
  • Strong acrylic adhesive works best on flat, smooth surfaces
  • Inkjet receptive polyester material
  • Designed for indoor use
Inkjet-receptive labels allows label creation with variable data and colour components in-house. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge