Brady® Polyester Label Tape for BradyJet™ J7300 Colour Label Printer
Lieferant: Brady
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
170452 176577 176624 176625 176600 176578
BRDY170452EA 522.06 EUR InStock
BRDY170452 BRDY176577 BRDY176624 BRDY176625 BRDY176600 BRDY176578
Brady® Polyester Label Tape for BradyJet™ J7300 Colour Label Printer
Etiketten
General-purpose polyester labels from Brady feature a specially formulated topcoat for improved durability, colour, and appearance.
  • Inkjet-receptive polyester material showcases vibrant colours, sharp text, and precise graphics
  • Enhanced topcoat works with durable J7300 inks to protect printed text and graphics from smudging and harsh conditions
  • Aggressive adhesive
  • Designed for indoor/outdoor use
Inkjet-receptive polyester labels adhere strongly to smooth surfaces for a wide range of applications, and are durable up to two years outdoors, even longer indoors. Ideal for Lean/5S, general facility ID, pipe marking, OSHA/ANSI labelling, machine labels, PPE and instructional signage, diagrams, quality and control labelling, and lockout tagout identification. Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 Colour Label Printer. Ink cartridges are required to print on this material; use part numbers P-CMY-1 and P-K-1.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge