EasySep™ Multistand
Lieferant: STEMCELL Technologies
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
EasySep™
STMC18010EA 2160 EUR InStock
STMC18010
EasySep™ Multistand
Magnete
The EasySep™ Multistand allows the separation of up to four samples at one time.
  • Immunomagnetic column-free magnet
An ingenious reversible clamping bar allows the multistand to hold up to 4 EasySep™ magnets or 4 EasySep™ 'The Big Easy' magnets. After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into receiving tubes. The receiving tube bar is indexed to ensure proper position of the tubes.
Bestellinformation: Magnets must be purchased separately.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge