Handheld Multifunction Meters, PRO D01, D05.2 and D05.3
Lieferant: Greisinger
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
486134 486137 486136
GRSG486134EA 0 EUR InStock
GRSG486134 GRSG486137 GRSG486136
Handheld Multifunction Meters, PRO D01, D05.2 and D05.3
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
PRO D01, D05.2 and D05.3 are a high class professional multifunction handheld meter with a rich set of features, high grade robustness and operating comfort for safe and reliable use. PRO D05.2 and PRO D05.3 also have data logging capabilities and a USB-rechargeable battery system.
  • PRO D01 has only one connector, PRO D05.2 has two connectors and PRO D05.3 has three connectors
  • Wide range of interchangeable and automatically recognised digital probes can be connected
  • Fast and accurate
  • Easy to use and read due to backlit dot matrix/clear text display
  • Various measurement views available, including life chart
  • Acoustic alarm with high/low thresholds and optional hysteresis
  • List of favorite functions for quick access to the most used operations
  • NiMH batteries rechargeable via USB
Statistical functions minimum, average, maximum. It has a built-in foldable stand and magnet for flexible operation. Shock and impact proof. IP 67 waterproof (except pressure ports).
Automatic with manual or scheduled start/stop logging type and the logging intervals are 1, 5, 10, 15, 30 s/1, 2, 5, 10, 15, 20, 30 min/one hour.
Bestellinformation: Sensors have to be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge