Spatel, rund, gebogen
Lieferant: BOCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
3200 3202 3203 3204 32005 3201 32025
231-1045EA 2.7 EUR InStock
231-1045 231-1047 BOCH3203 BOCH3204 BOCH32005 BOCH3201 BOCH32025
Spatel, rund, gebogen
Spatel
Stainless steel ¹⁸/₁₀, anti-magnetic. These spatulas with one rounded end can be used for a variety of applications.
  • Excellent heat resistance, autoclavable
  • Good resistance to chemicals
  • Easy and quick cleaning
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge