Schraubverschlüsse, PPCO, 13 mm
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
342826-0110 342826-0111 342826-0114
215-1584EA 205 EUR InStock
215-1584 215-1585 215-1586
Schraubverschlüsse, PPCO, 13 mm
Verschlüsse Chromatographieampullen-Verschlüsse
Identify samples quickly with micro packaging vial closures for 4,5 ml vials, offered in green, white or natural. Polypropylene polymer closures are ready to use and have no O-ring to fall out or contaminate contents.
  • RNase/DNase free (only sterile closures), non-cytotoxic and non-pyrogenic
Zertifizierungen: Manufactured in compliance with ISO 13485:2003 quality requirements. Micro packaging vials and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Verpackung: Closures are bulk packed without vials, 500 per inner bag, 1000 per case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge