Hei-FLOW Silver 1, Hei-FLOW Silver 2 Peristaltic Pump Packages
Lieferant: Heidolph Instruments GmbH & Co.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
523-50019-00 523-50019-02 523-50068-00 523-50068-02 523-50068-03
224-1455EA 3498 EUR InStock
224-1455 224-1456 224-0032 224-0040 HEID523-50068-03
Hei-FLOW Silver 1, Hei-FLOW Silver 2 Peristaltic Pump Packages
Pumpen Peristaltikpumpen
Packages for Hei-FLOW Value 01 Hei-FLOW Value 06 peristaltic pumps.
  • For standard applications and liquid transfer
Hei-FLOW Silver 1 Package includes Hei-FLOW Value 01 (model PD 5001), Pump head SP quick 1,6, and 1 m each Tygon® (standard) and Silicone tubing (inner Ø 3,1 mm).
Hei-FLOW Silver 2 Package includes Hei-FLOW Value 06 (model PD 5006), Pump head SP standard 2,5, and 1 m each Tygon® (standard) and Silicone tubing (inner Ø 6,4 mm).
Bestellinformation: For more information, see peristaltic pump drive models PD 5001 and PD 5006.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge