Erlenmeyerkolben, Weithals, mit Schraubverschluss
Lieferant: Brand
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
93170 93172 93175 93180 93185
BRND93170EA 34.2 EUR InStock
BRND93170 BRND93172 BRND93175 BRND93180 BRND93185
Erlenmeyerkolben, Weithals, mit Schraubverschluss
Kolben Erlenmeyerkolben
PP, highly translucent.
  • Very good chemical resistance
  • Resistant to heat up to +121 °C (for 20 m), prolonged use +100 °C
  • Also sealable with standard ground joint stoppers (stoppers not supplied)
With easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge