A10821.0B
A10821.0I
A10821.30
A10821.0BEA
152
EUR
InStock
A10821.0B
A10821.0I
A10821.30
1-Brompentan ≥99%
1-Brompentan
Purity: 99%
CAS No.: 110-53-2
Molecular Formula: CH3(CH2)4Br
Formula Weight: 151.05
Flash Point: 31°(87°F)
Refractive Index: 1.4436
Odour: Ether-like
Melting Point: -95°
Boiling Point: 127-129°
Beilstein Reference: 1730981
Merck Reference: 14,602
UN No.: 1993
MDL No.: MFCD00000267