L08457.03
L08457.06
L08457.03EA
39.9
EUR
InStock
L08457.03
L08457.06
3,5-Dihydroxybenzamid ≥97%
3,5-Dihydroxybenzamid
Purity: 97%
CAS No.: 3147-62-4
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Melting Point: ca 280° dec.
Beilstein Reference: 2690560