B24900.14
B24900.22
B24900.36
B24900.14EA
25
EUR
InStock
B24900.14
B24900.22
B24900.36
1-Chloroctadecan ≥94%
1-Chloroctadecan
Purity: 94%
CAS No.: 3386-33-2
Molecular Formula: C18H37Cl
Formula Weight: 288.95
Flash Point: 110°(230°F)
Refractive Index: 1.4510
Melting Point: 20-23°
Boiling Point: 157-158°/1.5mm
MDL No.: MFCD00000960