B21633.06
B21633.14
B21633.06EA
24.6
EUR
InStock
B21633.06
B21633.14
2H,3H-Perfluorpentan tech. 90%, technische Qualität
2H,3H-Perfluorpentan
Purity: 90%
CAS No.: 138495-42-8
Molecular Formula: C5H2F10
Formula Weight: 252.05
Flash Point: None
Refractive Index: 1.3000
Melting Point: -80°
Boiling Point: 54°
MDL No.: MFCD00798129