Spritzflaschen, enghalsig
Lieferant: KARTELL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
680 681 682
215-1365EA 9.2 EUR InStock
215-1365 215-1366 215-1367
Spritzflaschen, enghalsig
Flaschen Spritzflaschen
LDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Suitable for foodstuff
The stream of liquid is obtained by blowing through the straight tube. The direction of the stream of liquid can be controlled by hand.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge