L17024.03
L17024.06
L17024.03EA
42.3
EUR
InStock
L17024.03
L17024.06
(S)-(-)-1-(2-Naphthyl)ethylamin ≥99%, ee 99%, ChiPros®
(S)-(-)-1-(2-Naphthyl)ethylamin
Purity: 99+%
CAS No.: 3082-62-0
Molecular Formula: C12H13N
Formula Weight: 171.24
Melting Point: 48-52°
Boiling Point: 90°/0.15mm
UN No.: 3259
MDL No.: MFCD00085366