13406.30
13406.A1
13406.18
13406.30EA
110
EUR
InStock
13406.30
13406.A1
13406.18
Lithiumfluorid ≥98,5%
Lithium fluoride
Purity: 98.5%
CAS No.: 7789-24-4
Molecular Formula: LiF
Formula Weight: 25.94
Physical Form: -325 Mesh Powder
Density: 2.635
Refractive Index: 1.3915
Odour: Odourless
Melting Point: 845°
Boiling Point: 1680°
Solubility: Soluble in water and in acids
Merck Reference: 14,5531
UN No.: 3288
MDL No.: MFCD00011090
In vitreous enamels and glazes, as flux in soldering and welding aluminum, infrared spectrophotometers