Vibration spatulas, micro-blade, Quaverette®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
H367180000 H367190000
231-2417EA 180 EUR InStock
231-2417 BELAH367190000
Vibration spatulas, micro-blade, Quaverette®, SP Bel-Art
Spatel
Stainless steel or stainless steel with Teflon® FEP coating. The micro V-shaped narrow blade can be moved easily inside the small mouth containers and tight corners. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving control over transfer from spatula to weighing dish.
  • Reduces transfer losses
  • Make accurate additions of precious materials as needed
Zertifizierungen: Coating complies with FDA regulations governing direct food contact applications.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge