Magnetic hotplate stirrers, Undergrad UC152/D and US152/D
Lieferant: Cole Parmer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
Stuart®
STUAUC152D/KITEA 921 EUR InStock
STUAUC152D/KIT 442-1075
Magnetic hotplate stirrers, Undergrad UC152/D and US152/D
Rührer Magnetrührer mit Heizplatte
Compact general purpose hotplate stirrers designed for safety and performance. The hotplate has an innovative LED temperature indicator scale and can also be used in conjunction with the SCT1 digital contact thermometer to accurately control sample temperature. Powerful magnets and motor give stirring speeds up to 2000 min⁻¹ and unit is capable of mixing large volumes (up to 15 litres*). Both models have an integral fitting for a retort rod.
  • Choice of top plate: Robust coated aluminium or chemically resistant ceramic
  • Flashing 'hot' warning light whenever the plate temperature is above 50 °C, is mains independent so stays on even when the unit is unplugged
  • Accurate temperature control with LED temperature indicator on UC152 & US152
  • Digital display of hotplate temperature on UC152D & US152D
  • Compact, space saving units, can be stored on their side to maximise storage space
  • Designed with recess at the base for a retort stand to slide underneath unit, saves space and time when setting up
Model UC152/UC152D has a glass ceramic top which has excellent chemical resistance. The surface is easy to clean and the thermal properties allow very high plate temperatures while ensuring the edges stay cooler, reducing the chance of accidental burns. The white surface ensures good visibility of colour changes.
Model US152/US152D has a robust aluminium/silicon alloy top plate for excellent heat transmission. The top plate has a thin ceramic coating for added chemical resistance. A 700 W element gives rapid heating and ensures even temperature distribution across the whole surface of the plate.
Zertifizierungen: IP protection class according to DIN EN 60529: IP 32
Lieferumfang: Supplied complete with 2×25 mm PTFE coated magnetic stirring bars.
Achtung! Bitte beachten: * Based on water in a 20 litre glass bottom boiling flask.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge