Sicherheitswinterstiefel, zum Schnüren, Snow and Rain, Anatomic Bau 840 XP
Lieferant: ATLAS - the shoe company
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
06336 06337 06338 06339 06340 06341 06342 06343 06344 06345 06346 06347 06348 06349 06350
ATLA06336EA 0 EUR InStock
ATLA06336 ATLA06337 ATLA06338 ATLA06339 ATLA06340 ATLA06341 ATLA06342 ATLA06343 ATLA06344 ATLA06345 ATLA06346 ATLA06347 ATLA06348 ATLA06349 ATLA06350
Sicherheitswinterstiefel, zum Schnüren, Snow and Rain, Anatomic Bau 840 XP
Schuhe und Stiefel
Safety ankle boot made from waterproof leather, woven fur lining and a cold isolating ICE AGE outdoor® sole (4,5 mm rough tread profile, oil, acid and petrol resistant).
  • Isolating supporting surface and warm woven fur lining
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
  • Puncture resistant, metal-free XP® midsole
  • Large stud profile
  • Reinforced toe protection
Farbe: Schwarz
Zertifizierungen: DGUV 112-191; EN ISO 20345; S3, CI, SRC
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge