Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Split-hinged tube furnaces, for vertical operation, VST and TVS

Lieferant: CARBOLITE

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

TVS12600301 TVS126003216P1 TVS126003508P1 TVS12900301 TVS129003216P1 TVS129003508P1 VST12200301 VST122003216P1 VST122003508P1 VST12400301 VST124003216P1 VST124003508P1 VST12600301 VST126003216P1 VST126003508P1 VST12900301 VST129003216P1 VST129003508P1 TVS12/600/2604 VST120600230SN VST120900230SN
CARBTVS12600301EA 8330 EUR InStock
CARBTVS12600301 CARBTVS126003216P1 CARBTVS126003508P1 CARBTVS12900301 CARBTVS129003216P1 CARBTVS129003508P1 CARBVST12200301 CARBVST122003216P1 CARBVST122003508P1 CARBVST12400301 CARBVST124003216P1 CARBVST124003508P1 CARBVST12600301 CARBVST126003216P1 CARBVST126003508P1 CARBVST12900301 CARBVST129003216P1 CARBVST129003508P1 CARBTVS12/600/2604 CARBVST120600230SN CARBVST120900230SN
Split-hinged tube furnaces, for vertical operation, VST and TVS
Öfen Rohröfen
The vertical split tube furnace range VST (single zone) and TVS (three zone) uses free radiating wire elements embedded within the insulation of the furnace body. The benefit of this design is its flexibility with the use of tube adapters the same furnace can be used with a variety of tube diameters. These furnaces comprise a furnace body which is hinged and split into two halves along its length. This makes exchange of work tubes easier and also enables the furnace to be used with reactors or work tubes where end flanges would make insertion into a non-split furnace difficult. This design is particularly suitable for incorporation into extension test and creep test apparatus.

  • Maximum operating temperature 1200 °C
  • Carbolite Gero 301 controller, with single ramp to set-point & process timer
  • Accepts worktubes with outer diameters up to 110 mm
  • Vertical furnace with a separate control module on a 2 m conduit
  • Wire elements in high quality vacuum formed insulation ensure fast heat up, excellent temperature uniformity and short cool down times
  • Supplied in 'near-hinge' configuration complete with stand

Insulating end vestibules improve chamber uniformity and incorporate interchangeable tube adapters. Carbolite hinged tube furnaces are designed for bench mounting, and are interconnected to a separate control cabinet with a 6 foot cable. Positive break furnace safety switch isolates power to the heating elements when furnace is opened. Standard vertical hinged tube furnaces are provided with a vertical support stand.

Heated lengths: Single zone 200, 300, 400, 450, 600 or 900 mm (VST); 3-zone 600 or 900 mm (TVS)
End zone: Control is via back to back thermocouples (VST) or end zones of 150 mm each end (TVS)

Achtung! Bitte beachten: Note: This range of tube furnaces does not include an integral work tube and one must be selected as an additional item. The work tube length is dependent on the application eg for use with modified atmosphere or vacuum. The use of a separate work tube has the advantage of protecting the heating elements from damage or contamination.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge