Specialty vials
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
5180-0841 5180-0844 5182-3454 5183-2030 5183-2073 5183-4497 5183-4510 5184-3550 5184-3551 5184-3552 5184-3554 5184-3555 5184-3556
HEWL5180-0841EA 239 EUR InStock
HEWL5180-0841 HEWL5180-0844 HEWL5182-3454 HEWL5183-2030 HEWL5183-2073 HEWL5183-4497 HEWL5183-4510 HEWL5184-3550 HEWL5184-3551 HEWL5184-3552 HEWL5184-3554 HEWL5184-3555 HEWL5184-3556
Specialty vials
Vials
These speciality vials with wide opening are made of type I borosilicate glass.
High recovery glass vials are used for sample concentration and injection without transferring to microvolume inserts.
Micro-V vials are wineglass shaped used for microsampling.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge