PIG® spill kit, 76 L overpack drum
Lieferant: NEW PIG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
KIT211 KIT311 KITR311 KITR211
121-0053EA 223 EUR InStock
121-0053 121-0511 121-0512 121-0078
PIG® spill kit, 76 L overpack drum
Absorptionsmaterial
Strong enough to earn a UN rating for transporting spill cleanup waste.
  • Comply with spill plan regs, avoid fines and be ready to respond
  • Overpack drum container is UN-rated (UN1H2/X46/S) for transporting waste after spill cleanup
  • Neatly prepacked container speeds access during a spill emergency
  • Overpack is X-rated in Packing Groups I, II and III for transporting spill cleanup waste by land, sea or rail
  • Lightweight polypropylene container resists chemicals and keeps contents clean and dry
  • Two broom holes 180-degrees opposite each other on the lid allow 100% positive leverage for easy opening and closing with a broom handle or bar
The ledges molded into the container make kit easy to move by forklift; foot hold in the drum base allows you to hold the drum securely in position while opening or closing the lid.
Verpackung: In a protective insert with lid for long-term protection against UV degradation; contents are organised for quick spill response.
Achtung! Bitte beachten: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge